Learn More
Leica Biosystems METAL BSE MOLDS SUPR CASSETTES
Catalog No. NC1307541
Encompass_Preferred
Disposable Base Molds have been designed to be compatible with any currently available cassette. The orientation of the specimen is made easier due to the transparency of the mold, while its rounded corners allow for easy block removal and paraffin ribbon continuity. Built up side walls reduce risk of paraffin seepage during embedding. Available in boxes of 400 or cases of 1600.
Catalog No. NC1307541 Supplier Leica Biosystems Supplier No. 38VSP58166
Disposable Base Molds have been designed to be compatible with any currently available cassette. The orientation of the specimen is made easier due to the transparency of the mold, while its rounded corners allow for easy block removal and paraffin ribbon continuity. Built up side walls reduce risk of paraffin seepage during embedding. Available in boxes of 400 or cases of 1600.