Learn More
Fisherbrand™ Metal Cleanroom Binder
Catalog No. 18003116 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
1 in.
2 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
18-003-116 2 in.
18-003-115 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-003-116
Add to Cart
Add to Cart
Metal cleanroom binder, aluminum, 3 metal rings
  • Designed for Class 10 - Class 100,000 cleanrooms, our heavy-duty metal, 3-ring cleanroom binders are intended to be reused, not recycled.
  • The heavy-duty metal rings keep all cleanroom documents secure and in place. If dropped, you can be sure all documents will remain intact within the binder.
  • The ring release mechanism allows for easy placement and movement of documents, and locks back into place securely.
  • This item does not include poly pockets. It is intended to be used in sterile and non-sterile controlled environments.
  • Our metal cleanroom binders are auto- clavable-safe.
  • All our Class 10 binders are the industry standard and proudly made in America.
  • 75-gauge aluminum front cover, spine, and back cover
  • 200 sheet binder capacity for 1 in. binder
  • 400 sheet binder capacity for 2 in. binder
  • Each binder is individually double bagged
  • Autoclavable-safe
Specifications
Material Plastic
Color Black
Length (English) 2 in.
Product Type Cleanroom Binder