Learn More
Andwin Scientific METAL COOLING CHAMBERS F/0.2ML
SDP
Catalog No. NC9762282
Encompass_Preferred
This cooling chamber block features a solid aluminum construction designed to surround 0.2ml PCR plates or tubes to guarantee sustained hot or cold temperatures.
Catalog No. NC9762282 Supplier Andwin Scientific Supplier No. CHAM1000
This cooling chamber block features a solid aluminum construction designed to surround 0.2ml PCR plates or tubes to guarantee sustained hot or cold temperatures.