Learn More
MSC Vikan Shovel, Metal Detectable, Polypropylene, 10.2 in Width, White
Catalog No. 502836331
Encompass
This one-piece polypropylene shovel is tough, lightweight, and hygienic, molded from FDA-compliant polypropylene. It is suitable for moving bulk materials and includes a semi-ferrous additive for metal detectability, making it ideal for use with metal detectable systems. This shovel is popular in food handling applications such as meat, dairy, seafood, and poultry processing.
  • One-piece construction for enhanced hygiene and durability.
  • Metal detectable design suitable for metal detectable systems.
  • Made from FDA-compliant polypropylene.
  • Tough, lightweight, and hygienic properties.
  • Autoclavable for sterilization.
  • Halal and Kosher certified.
  • D-grip handle provides a secure grip.
  • Heavy-duty construction.
Catalog No. 50-283-6331 Supplier MSC Supplier No. 47170220
This one-piece polypropylene shovel is tough, lightweight, and hygienic, molded from FDA-compliant polypropylene. It is suitable for moving bulk materials and includes a semi-ferrous additive for metal detectability, making it ideal for use with metal detectable systems. This shovel is popular in food handling applications such as meat, dairy, seafood, and poultry processing.
  • One-piece construction for enhanced hygiene and durability.
  • Metal detectable design suitable for metal detectable systems.
  • Made from FDA-compliant polypropylene.
  • Tough, lightweight, and hygienic properties.
  • Autoclavable for sterilization.
  • Halal and Kosher certified.
  • D-grip handle provides a secure grip.
  • Heavy-duty construction.