Learn More
MSC Vikan Shovel, Polypropylene, 14 in Blade, Metal Detectable, White
Catalog No. 502839017
Encompass
This one-piece shovel is designed for transferring meat and dairy in food processing plants. Its seamless polypropylene construction offers durability, resists bacteria growth, and complies with FDA requirements. It is dishwasher and autoclave safe for easy cleaning and is lightweight to help prevent worker fatigue, with an overall length of 42.5 inches for ideal lifting leverage.
  • Seamless construction
  • FDA compliant
  • Lightweight design
  • Dishwasher and autoclave safe
  • Resists bacteria growth
  • Chemical-resistant
  • Corrosion-resistant
  • Hygienic construction
  • One-piece design
  • D-handle for ergonomic grip
  • Metal detectable for food safety
Catalog No. 50-283-9017 Supplier MSC Supplier No. 83850826
This one-piece shovel is designed for transferring meat and dairy in food processing plants. Its seamless polypropylene construction offers durability, resists bacteria growth, and complies with FDA requirements. It is dishwasher and autoclave safe for easy cleaning and is lightweight to help prevent worker fatigue, with an overall length of 42.5 inches for ideal lifting leverage.
  • Seamless construction
  • FDA compliant
  • Lightweight design
  • Dishwasher and autoclave safe
  • Resists bacteria growth
  • Chemical-resistant
  • Corrosion-resistant
  • Hygienic construction
  • One-piece design
  • D-handle for ergonomic grip
  • Metal detectable for food safety