Learn More
MSC Vikan One-Piece Metal Detectable Shovel, 13.7 inch Blade, Green, Polypropylene
Catalog No. 502838149
Encompass
This one-piece polypropylene shovel is tough, lightweight, and hygienic, molded from FDA-compliant material. Designed for moving bulk materials, it incorporates a semi-ferrous additive for metal detectable systems, offering a cost-effective alternative to stainless steel and helping reduce user fatigue. It is widely used in various food handling applications, including processing meat, dairy, seafood, and poultry, as well as commercial fishing.
  • Designed for critical hygiene and food safety environments.
  • Ergonomic D-grip and large pan for efficient bulk material handling.
  • Metal and x-ray detectable with food contact compliant semi-ferrous additives.
  • One-piece polypropylene construction enhances durability and hygiene.
  • Lightweight design reduces user fatigue.
  • Easy to clean and maintain, suitable for diverse food processing and handling applications.
Catalog No. 50-283-8149 Supplier MSC Supplier No. 47170493
This one-piece polypropylene shovel is tough, lightweight, and hygienic, molded from FDA-compliant material. Designed for moving bulk materials, it incorporates a semi-ferrous additive for metal detectable systems, offering a cost-effective alternative to stainless steel and helping reduce user fatigue. It is widely used in various food handling applications, including processing meat, dairy, seafood, and poultry, as well as commercial fishing.
  • Designed for critical hygiene and food safety environments.
  • Ergonomic D-grip and large pan for efficient bulk material handling.
  • Metal and x-ray detectable with food contact compliant semi-ferrous additives.
  • One-piece polypropylene construction enhances durability and hygiene.
  • Lightweight design reduces user fatigue.
  • Easy to clean and maintain, suitable for diverse food processing and handling applications.