Learn More
MSC Vikan Mixing Paddle, Metal Detectable, 52 in, Polypropylene, Blue
Catalog No. 502837260
Encompass
This durable Vikan mixing paddle is designed for food processing environments. Constructed from flexible polypropylene with a semi-ferrous additive, it is metal detectable, ensuring product safety. The paddle features a thin, tapered edge for efficient scraping of containers and is suitable for mixing various food products. Its one-piece design prevents dirt or germ harborage, and the blue color supports HACCP color-coded systems.
  • Color coded for HACCP systems
  • Dishwasher safe
  • Ergonomic handles for comfort and control
  • Manufactured in the United States
  • Metal detectable with a semi-ferrous additive
  • Autoclave safe for sterilization
  • Flexible blade with tapered edge for efficient scraping
  • Long handle to maintain hygiene
  • Lightweight design to reduce user fatigue
  • Withstands temperatures up to 266 degrees F
Catalog No. 50-283-7260 Supplier MSC Supplier No. 44930709
This durable Vikan mixing paddle is designed for food processing environments. Constructed from flexible polypropylene with a semi-ferrous additive, it is metal detectable, ensuring product safety. The paddle features a thin, tapered edge for efficient scraping of containers and is suitable for mixing various food products. Its one-piece design prevents dirt or germ harborage, and the blue color supports HACCP color-coded systems.
  • Color coded for HACCP systems
  • Dishwasher safe
  • Ergonomic handles for comfort and control
  • Manufactured in the United States
  • Metal detectable with a semi-ferrous additive
  • Autoclave safe for sterilization
  • Flexible blade with tapered edge for efficient scraping
  • Long handle to maintain hygiene
  • Lightweight design to reduce user fatigue
  • Withstands temperatures up to 266 degrees F