Learn More
MSC Vikan Mixing Paddle, Metal Detectable, 52 in, Red
Catalog No. 502835636
Encompass
This mixing paddle is designed for use in environments where metal detection is critical. Constructed from polypropylene with a semi-ferrous additive, it is metal detectable, helping to prevent contamination. The paddle is 52 inches long and features a solid design without holes, making it suitable for a variety of mixing tasks.
  • Metal detectable material for enhanced food safety
  • Constructed from durable polypropylene
  • Designed without holes for versatile mixing applications
  • Originating from the United States
Catalog No. 50-283-5636 Supplier MSC Supplier No. 44930758
This mixing paddle is designed for use in environments where metal detection is critical. Constructed from polypropylene with a semi-ferrous additive, it is metal detectable, helping to prevent contamination. The paddle is 52 inches long and features a solid design without holes, making it suitable for a variety of mixing tasks.
  • Metal detectable material for enhanced food safety
  • Constructed from durable polypropylene
  • Designed without holes for versatile mixing applications
  • Originating from the United States