Learn More
Cole-Parmer Digestion Cup, PFA with PFA Cap, 50 mL; 1/Ea
Supplier:  Cole-Parmer 5320202
Encompass
Savillex SC545 Digestion Cup, PFA with PFA Cap, 50 mL; 1/Ea, Disposable digestion cups eliminate transfer stepsCups that act as a digestion vessel or storage container and fit perfectly into the HotBlock and HotBlock Pro systemsLow metals background and EPA approved for digestionsAccessories include watch glasses, reflux caps, and transfer/storage racksCompatible with 50 mL HotBlock Systemsnot compatible with FlipMate
Catalog No. NC1076786