Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Metal Dual Plate Holder Assembly for FastPrep-96™

Catalog No. MP119696169 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
MP119696169 FastPrep-96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119696169 Supplier MP Biomedicals™ Supplier No. 119696169
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended for high-throughput samples and can hold deep well plates.

  • Adapter for FastPrep-96™ & 96 Pro™
  • Holds 2 x 96 deep well plates
  • Anodized aluminum metal
  • Wide temperature range resistance
TRUSTED_SUSTAINABILITY

Specifications

Product Type Dual Plate Holder
Blade Material Stainless Steel
Closure Color Blue
For Use With (Equipment) FastPrep-96

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.