Learn More
Med Vet International Miltex Vantage Metal Ear Syringe, 4oz. Capacity
Catalog No. 502830260
Encompass_Preferred
The Miltex Vantage V919-385 ear / ulcer bulb syringe has a 4 ounce (118ml) capacity. It is chrome-plated and without gradations.
Catalog No. 50-283-0260 Supplier Med Vet International Supplier No. MILV919385
The Miltex Vantage V919-385 ear / ulcer bulb syringe has a 4 ounce (118ml) capacity. It is chrome-plated and without gradations.