Learn More
Hach Company Radiometer Analytical MC3051Pt Combination Metal Electrode (Ag/AgCl reference, platinum ring sensor, screw cap)
Catalog No. 501997597 Shop All Hach Company Products
Encompass
The Radiometer MC3051Pt Combination Metal Electrode has a Ag/AgCl reference system with refillable electrolyte (3 M KCl + sat. AgCl), porous pin junction, and platinum ring sensor. The MC3051Pt electrode is recommended for general redox measurements. The MC3051Pt is a screw cap electrode and requires the CL114 cable (part no. A94L114) for use with Hach Sension+ laboratory and other BNC-compatible meters/titrators.
Catalog No. 50-199-7597 Supplier Hach Company Supplier No. E31M003
The Radiometer MC3051Pt Combination Metal Electrode has a Ag/AgCl reference system with refillable electrolyte (3 M KCl + sat. AgCl), porous pin junction, and platinum ring sensor. The MC3051Pt electrode is recommended for general redox measurements. The MC3051Pt is a screw cap electrode and requires the CL114 cable (part no. A94L114) for use with Hach Sension+ laboratory and other BNC-compatible meters/titrators.