Learn More
Hach Company Radiometer Analytical XM150 Metal Electrode (for Voltammetry applications, platinum disc sensor, screw cap)
Catalog No. 501997540 Shop All Hach Company Products
Encompass
The Radiometer XM150 Metal Electrode has a platinum disc sensor and is recommended for Voltammetry applications. The XM150 is a screw cap electrode and requires an external connection cable for connection to a meter.
Catalog No. 50-199-7540 Supplier Hach Company Supplier No. B35M150
The Radiometer XM150 Metal Electrode has a platinum disc sensor and is recommended for Voltammetry applications. The XM150 is a screw cap electrode and requires an external connection cable for connection to a meter.