Learn More
Hach Company Radiometer Analytical XM120 Metal Electrode (for Voltammetry applications, platinum plate sensor, screw cap)
Catalog No. 501997538 Shop All Hach Company Products
Encompass
The Radiometer XM120 Metal Electrode has a platinum plate sensor and is recommended for Voltammetry applications. The XM120 is a screw cap electrode and requires an external connection cable for connection to a meter.
Catalog No. 50-199-7538 Supplier Hach Company Supplier No. B35M120
The Radiometer XM120 Metal Electrode has a platinum plate sensor and is recommended for Voltammetry applications. The XM120 is a screw cap electrode and requires an external connection cable for connection to a meter.