Learn More
Hach Company Radiometer Analytical M231Pt Metal Electrode (platinum wire sensor, screw cap)
Catalog No. 501997596 Shop All Hach Company Products
Encompass
The Radiometer M231Pt Metal Electrode has a platinum wire sensor and is recommended for general purpose measurements. An external reference electrode is required. The M231Pt is a screw cap electrode and requires the CL114 cable (part no. A94L114) for use with Hach Sension+ laboratory and other BNC-compatible meters/titrators.
Catalog No. 50-199-7596 Supplier Hach Company Supplier No. E31M002
The Radiometer M231Pt Metal Electrode has a platinum wire sensor and is recommended for general purpose measurements. An external reference electrode is required. The M231Pt is a screw cap electrode and requires the CL114 cable (part no. A94L114) for use with Hach Sension+ laboratory and other BNC-compatible meters/titrators.