Learn More
World Precision Instrument Metal Electrodes Assortment Package, TM53A05KT (3), TM53A10KT (3), TM53A20KT (4), Profile C
SDP
Catalog No. 502538432
Encompass
Introductory Assortments
Each of these assortment kits includes electrodes with different impedance within each style. Use an assortment kit to determine which electrode you need for your experiment. Ten electrodes per box, no mixing
Contains the following electrode impedances by quantity (package of 5)
TM53A05KT (3), TM53A10KT (3), TM53A20KT (4)
Frequently Asked Questions
Catalog No. 50-253-8432 Supplier World Precision Instrument Supplier No. TM53AXXKT
Introductory Assortments
Each of these assortment kits includes electrodes with different impedance within each style. Use an assortment kit to determine which electrode you need for your experiment. Ten electrodes per box, no mixing
Contains the following electrode impedances by quantity (package of 5)
TM53A05KT (3), TM53A10KT (3), TM53A20KT (4)
Frequently Asked Questions