Learn More
MSC METAL ENERGY ABSORBER KIT
Catalog No. 5000793927
Encompass
This Energy Absorber Kit is designed for integration into horizontal lifeline systems, providing mobility and fall protection when working at height. It includes essential components like an energy absorber, shackles, and tensioners to simplify system assembly and enhance safety by limiting fall arrest forces.
  • For use with temporary horizontal lifeline systems
  • Limits fall arrest forces to safe levels
  • Allows use of standard 5,000 lb. (22.2 kN) anchors
  • Configurable for system lengths up to 60 ft. (18.3 m) with one device or 100 ft. (30.5 m) with two
  • Lightweight and portable design
  • Extremely durable aluminum construction
  • Supports a maximum of 2 workers
  • Weight capacity of 310 lb. (141 kg)
Catalog No. 50-007-93927 Supplier MSC Supplier No. 71067706
This Energy Absorber Kit is designed for integration into horizontal lifeline systems, providing mobility and fall protection when working at height. It includes essential components like an energy absorber, shackles, and tensioners to simplify system assembly and enhance safety by limiting fall arrest forces.
  • For use with temporary horizontal lifeline systems
  • Limits fall arrest forces to safe levels
  • Allows use of standard 5,000 lb. (22.2 kN) anchors
  • Configurable for system lengths up to 60 ft. (18.3 m) with one device or 100 ft. (30.5 m) with two
  • Lightweight and portable design
  • Extremely durable aluminum construction
  • Supports a maximum of 2 workers
  • Weight capacity of 310 lb. (141 kg)