Learn More
Mopec Metal Evidence Containers
SDP
Catalog No. 22446034 Shop All Mopec Products
Change view
Click to view available options
Quantity:
0.5 oz.
1 oz.
2 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-446-034 1 oz.
22-446-026 0.5 oz.
22-446-027 2 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-446-034 Supplier Mopec Supplier No. FS319
Add to Cart
Add to Cart
Designed to protect small, fragile items like bullets, shell casings, and glass shards while supporting secure sealing and identification.
  • Features a Write-on label for easy identification.
  • Can be securely sealed with evidence tape or seals.
Specifications
Product Type Evidence Container
Color Silver with White Label
Quantity 1 oz.