Learn More
Grainger Metal Hose 1-1/2 ID x 10 ft.L
Catalog No. 502763867
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1-1/2" diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made of 304 stainless steel with a smooth inner wall. The hose has a temperature range of -40°F to 1000°F. It comes in a 25 ft length and has male NPT threaded ends. The hose is designed for use in industrial and commercial applications to convey air, water, oil and other fluids. It offers flexibility, durability and resistance to kinks. The interlocked corrugations allow it to be bent without compromising flow.
Catalog No. 50-276-3867 Supplier Grainger Supplier No. 16T826
The Hose Master Interlocked Metal Hose is a 1-1/2" diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made of 304 stainless steel with a smooth inner wall. The hose has a temperature range of -40°F to 1000°F. It comes in a 25 ft length and has male NPT threaded ends. The hose is designed for use in industrial and commercial applications to convey air, water, oil and other fluids. It offers flexibility, durability and resistance to kinks. The interlocked corrugations allow it to be bent without compromising flow.