Learn More
Grainger Metal Hose 1-1/2 ID x 12 ft.L
Catalog No. 502764035
Encompass_Preferred
The Hose Master Interlocked Metal Hose 1-1/2 in x 25 ft is a flexible metal hose with interlocking annular corrugations that allow it to be bent and shaped while maintaining strength and durability. It is made from 304 stainless steel with a working pressure rating of 300 PSI and temperature range of -40°F to 1000°F. This general purpose hose has a 1-1/2 inch inside diameter and is 25 feet in length. It has male NPT threaded ends and includes two swivel joint socket clamps for easy installation. The corrugated stainless steel construction makes it ideal for conveying air, water, oil and other fluids in industrial applications.
Catalog No. 50-276-4035 Supplier Grainger Supplier No. 16T827
The Hose Master Interlocked Metal Hose 1-1/2 in x 25 ft is a flexible metal hose with interlocking annular corrugations that allow it to be bent and shaped while maintaining strength and durability. It is made from 304 stainless steel with a working pressure rating of 300 PSI and temperature range of -40°F to 1000°F. This general purpose hose has a 1-1/2 inch inside diameter and is 25 feet in length. It has male NPT threaded ends and includes two swivel joint socket clamps for easy installation. The corrugated stainless steel construction makes it ideal for conveying air, water, oil and other fluids in industrial applications.