Learn More
Grainger Metal Hose 1-1/2 ID x 12 ft.L
Catalog No. 502767248
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 2 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a smooth inner wall and interlocked construction for flexibility. This hose has reusable fittings and is recommended for air, water, oil and other compatible fluids. It can withstand temperatures from -40 to 300 degrees Fahrenheit. The hose is sold by the foot and features a 1 year warranty from Hose Master.
Catalog No. 50-276-7248 Supplier Grainger Supplier No. 16T778
The Hose Master Interlocked Metal Hose is a 2 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a smooth inner wall and interlocked construction for flexibility. This hose has reusable fittings and is recommended for air, water, oil and other compatible fluids. It can withstand temperatures from -40 to 300 degrees Fahrenheit. The hose is sold by the foot and features a 1 year warranty from Hose Master.