Learn More
Grainger Metal Hose 1-1/2 ID x 14 ft.L
Catalog No. 502765127
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1" diameter x 25' long stainless steel metal hose with interlocking annular corrugations. It has a 300 psi working pressure and temperature range of -40°F to 1000°F. This flexible metal hose is made in the USA and has zinc-plated carbon steel fittings. It can be used for conveying air, water, oil and other fluids in industrial applications. The interlocked corrugations allow it to be bent and shaped while maintaining strength and leak resistance. It is durable and crush resistant.
Catalog No. 50-276-5127 Supplier Grainger Supplier No. 16T779
The Hose Master Interlocked Metal Hose is a 1" diameter x 25' long stainless steel metal hose with interlocking annular corrugations. It has a 300 psi working pressure and temperature range of -40°F to 1000°F. This flexible metal hose is made in the USA and has zinc-plated carbon steel fittings. It can be used for conveying air, water, oil and other fluids in industrial applications. The interlocked corrugations allow it to be bent and shaped while maintaining strength and leak resistance. It is durable and crush resistant.