Learn More
Grainger Metal Hose 1-1/2 ID x 16 ft.L
Catalog No. 502767352
Encompass_Preferred
The Hose Master 1LND8 is a flexible metal hose assembly with a 1" NPT male end and 1" NPT female swivel end. It has a working pressure of 300 PSI and a maximum temperature of 450°F. The corrugated stainless steel hose is 12" in length and covered with a stainless steel exterior braid. This durable, kink resistant hose provides flexibility for connecting rigid piping or equipment. It can be used for hydraulic oil, air, water, steam, petroleum products, and many chemicals. The assembly meets SAE 100R2 standards.
Catalog No. 50-276-7352 Supplier Grainger Supplier No. 16T780
The Hose Master 1LND8 is a flexible metal hose assembly with a 1" NPT male end and 1" NPT female swivel end. It has a working pressure of 300 PSI and a maximum temperature of 450°F. The corrugated stainless steel hose is 12" in length and covered with a stainless steel exterior braid. This durable, kink resistant hose provides flexibility for connecting rigid piping or equipment. It can be used for hydraulic oil, air, water, steam, petroleum products, and many chemicals. The assembly meets SAE 100R2 standards.