Learn More
Grainger Metal Hose 1-1/2 ID x 16 ft.L
Catalog No. 502764644
Encompass_Preferred
The Grainger 16" Stainless Steel Corrugated Metal Hose has a 1" ID and is made from 304 stainless steel. It has a working pressure of 150 PSI and temperature range of -425°F to 1000°F. The corrugated design allows flexibility. Common applications include connecting rigid piping systems that require vibration absorption or motion compensation. This high quality hose meets SAE J517 standard and is ideal for use with hot oils, water, steam, chemicals, petroleum products, and food products. The stainless steel material offers durability and corrosion resistance. With proper installation and care this hose will provide reliable performance.
Catalog No. 50-276-4644 Supplier Grainger Supplier No. 16T829
The Grainger 16" Stainless Steel Corrugated Metal Hose has a 1" ID and is made from 304 stainless steel. It has a working pressure of 150 PSI and temperature range of -425°F to 1000°F. The corrugated design allows flexibility. Common applications include connecting rigid piping systems that require vibration absorption or motion compensation. This high quality hose meets SAE J517 standard and is ideal for use with hot oils, water, steam, chemicals, petroleum products, and food products. The stainless steel material offers durability and corrosion resistance. With proper installation and care this hose will provide reliable performance.