Learn More
Grainger Metal Hose 1-1/2 ID x 18 ft.L
Catalog No. 502767282
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1-1/2" ID x 18' L interlocked metal hose with a working pressure of 300 PSI. It has a zinc-plated steel construction and is designed for use with hydraulic fluid, air, water, mild chemicals, and petroleum products. This flexible metal hose features a leak-proof design with no gaskets or seals. It connects using NPT threads and can handle temperatures from -40°F to 300°F. The interlocked construction allows it to be bent into tight radii without kinking.
Catalog No. 50-276-7282 Supplier Grainger Supplier No. 16T781
The Hose Master Interlocked Metal Hose is a 1-1/2" ID x 18' L interlocked metal hose with a working pressure of 300 PSI. It has a zinc-plated steel construction and is designed for use with hydraulic fluid, air, water, mild chemicals, and petroleum products. This flexible metal hose features a leak-proof design with no gaskets or seals. It connects using NPT threads and can handle temperatures from -40°F to 300°F. The interlocked construction allows it to be bent into tight radii without kinking.