Learn More
Grainger Metal Hose 1-1/2 ID x 18 ft.L
Catalog No. 502764007
Encompass_Preferred
The Hose Master Flexible Metal Hose Assembly is a stainless steel braided hose with brass fittings. It has a 1/4" NPT male inlet and outlet, is 12" in length, and rated for up to 5000 psi working pressure. The corrugated stainless steel inner tube is covered by a stainless steel wire braid reinforcement for strength and durability. This flexible metal hose can be used for hydraulic applications involving high pressure fluid transfer. The assembly is made in the USA.
Catalog No. 50-276-4007 Supplier Grainger Supplier No. 16T830
The Hose Master Flexible Metal Hose Assembly is a stainless steel braided hose with brass fittings. It has a 1/4" NPT male inlet and outlet, is 12" in length, and rated for up to 5000 psi working pressure. The corrugated stainless steel inner tube is covered by a stainless steel wire braid reinforcement for strength and durability. This flexible metal hose can be used for hydraulic applications involving high pressure fluid transfer. The assembly is made in the USA.