Learn More
Grainger Metal Hose 1-1/2 ID x 20 ft.L
Catalog No. 502764036
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1" diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a corrugated interlocked design for flexibility. This hose has a temperature range of -40°F to 1000°F and includes 316 stainless steel fittings. The hose is 25 feet long and has a 1" NPT male end and 1" NPT female swivel end. It can be used for conveying air, water, oil and other fluids in industrial applications. The interlocked corrugated design allows it to be bent without kinking.
Catalog No. 50-276-4036 Supplier Grainger Supplier No. 16T831
The Hose Master Interlocked Metal Hose is a 1" diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a corrugated interlocked design for flexibility. This hose has a temperature range of -40°F to 1000°F and includes 316 stainless steel fittings. The hose is 25 feet long and has a 1" NPT male end and 1" NPT female swivel end. It can be used for conveying air, water, oil and other fluids in industrial applications. The interlocked corrugated design allows it to be bent without kinking.