Learn More
Grainger Metal Hose 1-1/2 ID x 20 ft.L
Catalog No. 502765642
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1-1/2" diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel interlocked annular corrugated tubing and is designed for use with petroleum-based fluids including gasoline, diesel fuel, lubrication oils, hydraulic fluid and more. This metal hose offers flexibility while maintaining shape and form. It features reusable end fittings and is rated for temperatures up to 850°F. The hose has a minimum bend radius of 6" and is available in lengths up to 50 feet. It meets SAE J517 specifications.
Catalog No. 50-276-5642 Supplier Grainger Supplier No. 16T782
The Hose Master Interlocked Metal Hose is a 1-1/2" diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel interlocked annular corrugated tubing and is designed for use with petroleum-based fluids including gasoline, diesel fuel, lubrication oils, hydraulic fluid and more. This metal hose offers flexibility while maintaining shape and form. It features reusable end fittings and is rated for temperatures up to 850°F. The hose has a minimum bend radius of 6" and is available in lengths up to 50 feet. It meets SAE J517 specifications.