Learn More
Grainger Metal Hose 1-1/2 ID x 6 ft.L
Catalog No. 502764643
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1" diameter x 25' long metal hose with interlocked construction. It has a working pressure of 300 PSI and a burst pressure of 1200 PSI. The hose is made from 304 stainless steel with zinc-plated steel fittings. It has a temperature range of -40°F to 300°F. The hose is kink resistant and suitable for air, water, oil, and many chemicals. It connects using standard garden hose fittings. This durable metal hose is ideal for industrial, commercial, and residential applications.
Catalog No. 50-276-4643 Supplier Grainger Supplier No. 16T824
The Hose Master Interlocked Metal Hose is a 1" diameter x 25' long metal hose with interlocked construction. It has a working pressure of 300 PSI and a burst pressure of 1200 PSI. The hose is made from 304 stainless steel with zinc-plated steel fittings. It has a temperature range of -40°F to 300°F. The hose is kink resistant and suitable for air, water, oil, and many chemicals. It connects using standard garden hose fittings. This durable metal hose is ideal for industrial, commercial, and residential applications.