Learn More
Grainger Metal Hose 1-1/2 ID x 6 ft.L
Catalog No. 502765644
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1" diameter x 25' long metal hose with interlocking aluminum construction. It has a 300 psi working pressure and is designed for use with air, water, oil and other non-corrosive fluids. The interlocked aluminum provides flexibility while maintaining strength and shape. It has zinc-plated steel fittings with 1" NPT male threaded ends. The hose weighs 5 lbs and has a temperature range of -40°F to 300°F. It meets SAE 100R2 standards and is made in the USA.
Catalog No. 50-276-5644 Supplier Grainger Supplier No. 16T775
The Hose Master Interlocked Metal Hose is a 1" diameter x 25' long metal hose with interlocking aluminum construction. It has a 300 psi working pressure and is designed for use with air, water, oil and other non-corrosive fluids. The interlocked aluminum provides flexibility while maintaining strength and shape. It has zinc-plated steel fittings with 1" NPT male threaded ends. The hose weighs 5 lbs and has a temperature range of -40°F to 300°F. It meets SAE 100R2 standards and is made in the USA.