Learn More
Grainger Metal Hose 1-1/2 ID x 8 ft.L
Catalog No. 502767381
Encompass_Preferred
The Grainger 16T776 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and protected from dirt and moisture. This high quality bottle jack is designed for shop use and medium duty lifting applications.
Catalog No. 50-276-7381 Supplier Grainger Supplier No. 16T776
The Grainger 16T776 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and protected from dirt and moisture. This high quality bottle jack is designed for shop use and medium duty lifting applications.