Learn More
Grainger Metal Hose 1-1/2 ID x 8 ft.L
Catalog No. 502763965
Encompass_Preferred
The Grainger 16T825 is a Dayton 1/3 HP 115/230V AC electric motor with a totally enclosed fan cooled enclosure. It has a 1,725 RPM operating speed and features sleeve bearings. This single phase motor has a 48Y frame size and measures approximately 9-1/2" long by 5-3/4" wide. It requires a 48Y mount and has a 1/2" diameter shaft with a 1/4" keyway and set screw. The 16T825 operates on 115V AC and draws 8.6 amps, while on 230V AC it draws 4.3 amps. This electric motor is thermally protected for overloads.
Catalog No. 50-276-3965 Supplier Grainger Supplier No. 16T825
The Grainger 16T825 is a Dayton 1/3 HP 115/230V AC electric motor with a totally enclosed fan cooled enclosure. It has a 1,725 RPM operating speed and features sleeve bearings. This single phase motor has a 48Y frame size and measures approximately 9-1/2" long by 5-3/4" wide. It requires a 48Y mount and has a 1/2" diameter shaft with a 1/4" keyway and set screw. The 16T825 operates on 115V AC and draws 8.6 amps, while on 230V AC it draws 4.3 amps. This electric motor is thermally protected for overloads.