Learn More
Grainger Metal Hose 2-1/2 ID x 12 ft.L
Catalog No. 502765980
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It has a stainless steel interlock construction and is designed for use with petroleum-based fluids like diesel fuel, gasoline, and oil. The hose offers flexibility while maintaining strength and durability. It can withstand temperatures from -40 to 300 degrees Fahrenheit. The hose is 25 feet long and has male NPT ends. It meets SAE J517 specifications. This heavy duty metal hose is ideal for fuel transfer applications.
Catalog No. 50-276-5980 Supplier Grainger Supplier No. 16T836
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It has a stainless steel interlock construction and is designed for use with petroleum-based fluids like diesel fuel, gasoline, and oil. The hose offers flexibility while maintaining strength and durability. It can withstand temperatures from -40 to 300 degrees Fahrenheit. The hose is 25 feet long and has male NPT ends. It meets SAE J517 specifications. This heavy duty metal hose is ideal for fuel transfer applications.