Learn More
Grainger Metal Hose 2-1/2 ID x 16 ft.L
Catalog No. 502768302
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 6 inch diameter interlocked metal hose with a working pressure of 300 psi. It is made from 304 stainless steel with a smooth interior and exterior finish. The hose has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT threaded ends. The interlocked construction allows it to be flexible while still maintaining strength and durability. This stainless steel hose is ideal for conveying hot exhaust gases, fumes, and vapors.
Catalog No. 50-276-8302 Supplier Grainger Supplier No. 16T838
The Hose Master Interlocked Metal Hose is a 6 inch diameter interlocked metal hose with a working pressure of 300 psi. It is made from 304 stainless steel with a smooth interior and exterior finish. The hose has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT threaded ends. The interlocked construction allows it to be flexible while still maintaining strength and durability. This stainless steel hose is ideal for conveying hot exhaust gases, fumes, and vapors.