Learn More
Grainger Metal Hose 2-1/2 ID x 18 ft.L
Catalog No. 502766026
Encompass_Preferred
The Hose Master Flexible Metal Hose Assembly is a stainless steel flexible metal hose with a 1" NPT male end and a 1" NPT female swivel end. It has a working pressure of up to 300 PSI and a maximum temperature of 850°F. The corrugated stainless steel hose allows flexibility while maintaining strength and durability. It has a length of 12" and is made in the USA. The flexible metal hose is ideal for connecting rigid piping that requires flexibility and vibration absorption. Common applications include engine exhaust, fume removal, and transferring liquids, air, and gases in tight spaces.
Catalog No. 50-276-6026 Supplier Grainger Supplier No. 16T839
The Hose Master Flexible Metal Hose Assembly is a stainless steel flexible metal hose with a 1" NPT male end and a 1" NPT female swivel end. It has a working pressure of up to 300 PSI and a maximum temperature of 850°F. The corrugated stainless steel hose allows flexibility while maintaining strength and durability. It has a length of 12" and is made in the USA. The flexible metal hose is ideal for connecting rigid piping that requires flexibility and vibration absorption. Common applications include engine exhaust, fume removal, and transferring liquids, air, and gases in tight spaces.