Learn More
Grainger Metal Hose 2-1/2 ID x 20 ft.L
Catalog No. 502766117
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 2-1/2 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made of 304 stainless steel with a smooth inner wall. The hose has a temperature range of -40°F to 1000°F. It has a zinc-plated steel fitting on one end and is 36 inches long when fully extended. The hose allows for a 3:1 safety factor and is designed for use in hydraulic and pneumatic systems to absorb vibration and thermal expansion. It meets SAE 100R2 standards. The hose provides flexibility while maintaining strength and durability.
Catalog No. 50-276-6117 Supplier Grainger Supplier No. 16T840
The Hose Master Interlocked Metal Hose is a 2-1/2 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made of 304 stainless steel with a smooth inner wall. The hose has a temperature range of -40°F to 1000°F. It has a zinc-plated steel fitting on one end and is 36 inches long when fully extended. The hose allows for a 3:1 safety factor and is designed for use in hydraulic and pneumatic systems to absorb vibration and thermal expansion. It meets SAE 100R2 standards. The hose provides flexibility while maintaining strength and durability.