Learn More
Grainger Metal Hose 2-1/2 ID x 6 ft.L
Catalog No. 502765584
Encompass_Preferred
The Grainger 2-1/2" ID x 6' L Metal Hose is a flexible metal hose made from 304 stainless steel. It has a working pressure of up to 450 PSI and temperature range of -425°F to 1000°F. This hose features a corrugated design for flexibility and has a 2-1/2" inner diameter. It comes with a male NPT thread on each end and includes one swivel connector. The hose is rated for use with hydraulic oil, steam, water, air and inert gas. It meets SAE 100R2 standards. This metal hose would be useful for routing fluids and gases in industrial applications that require high working pressures and extreme temperature compatibility.
Catalog No. 50-276-5584 Supplier Grainger Supplier No. 16T833
The Grainger 2-1/2" ID x 6' L Metal Hose is a flexible metal hose made from 304 stainless steel. It has a working pressure of up to 450 PSI and temperature range of -425°F to 1000°F. This hose features a corrugated design for flexibility and has a 2-1/2" inner diameter. It comes with a male NPT thread on each end and includes one swivel connector. The hose is rated for use with hydraulic oil, steam, water, air and inert gas. It meets SAE 100R2 standards. This metal hose would be useful for routing fluids and gases in industrial applications that require high working pressures and extreme temperature compatibility.