Learn More
Grainger Metal Hose 2-1/2 ID x 8 ft.L
Catalog No. 502767526
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 2-1/2 inch diameter metal hose with a working pressure of 300 PSI. It has a stainless steel interlock construction with a smooth inner surface for efficient flow. The hose is designed for use with petroleum-based fluids like oil, diesel fuel, gasoline, and lubricants. It can withstand temperatures from -40 to 300°F. The metal hose is available in 25 ft, 50 ft, and 100 ft lengths and includes fittings on both ends. It meets SAE 100R2 standards for hydraulic hose.
Catalog No. 50-276-7526 Supplier Grainger Supplier No. 16T834
The Hose Master Interlocked Metal Hose is a 2-1/2 inch diameter metal hose with a working pressure of 300 PSI. It has a stainless steel interlock construction with a smooth inner surface for efficient flow. The hose is designed for use with petroleum-based fluids like oil, diesel fuel, gasoline, and lubricants. It can withstand temperatures from -40 to 300°F. The metal hose is available in 25 ft, 50 ft, and 100 ft lengths and includes fittings on both ends. It meets SAE 100R2 standards for hydraulic hose.