Learn More
Grainger Metal Hose 2 ID x 10 ft.L
Catalog No. 502767471
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a flexible metal hose that provides corrosion resistance and durability. It has a stainless steel interlock design that allows tight bending without kinking. This 1/4 inch hose has a working pressure of up to 1000 psi and a temperature range of -40°F to 1000°F. The interlocked stainless steel construction makes this hose ideal for applications where flexibility, corrosion resistance, and durability are needed. It can be used to convey air, water, oil and other fluids in harsh environments. This high quality metal hose from Hose Master is made in the USA.
Catalog No. 50-276-7471 Supplier Grainger Supplier No. 16T785
The Hose Master Interlocked Metal Hose is a flexible metal hose that provides corrosion resistance and durability. It has a stainless steel interlock design that allows tight bending without kinking. This 1/4 inch hose has a working pressure of up to 1000 psi and a temperature range of -40°F to 1000°F. The interlocked stainless steel construction makes this hose ideal for applications where flexibility, corrosion resistance, and durability are needed. It can be used to convey air, water, oil and other fluids in harsh environments. This high quality metal hose from Hose Master is made in the USA.