Learn More
Grainger Metal Hose 2 ID x 12 ft.L
Catalog No. 502765764
Encompass_Preferred
The PENFLEX Flexible Metal Hose Assembly is a stainless steel flexible hose with a 1" NPT male end connection on one side and a 1" NPT female swivel on the other side. It has a working pressure of up to 300 PSI and a maximum temperature of 1000°F. The flexible corrugated stainless steel construction allows it to absorb vibration and compensate for misalignment. With a 33" length, this flexible metal hose assembly is well-suited for connecting fixed piping systems to equipment when vibration absorption is required. The stainless steel material also offers corrosion resistance. This product can be used in a variety of industrial applications.
Catalog No. 50-276-5764 Supplier Grainger Supplier No. 16T786
The PENFLEX Flexible Metal Hose Assembly is a stainless steel flexible hose with a 1" NPT male end connection on one side and a 1" NPT female swivel on the other side. It has a working pressure of up to 300 PSI and a maximum temperature of 1000°F. The flexible corrugated stainless steel construction allows it to absorb vibration and compensate for misalignment. With a 33" length, this flexible metal hose assembly is well-suited for connecting fixed piping systems to equipment when vibration absorption is required. The stainless steel material also offers corrosion resistance. This product can be used in a variety of industrial applications.