Learn More
Grainger Metal Hose 2 ID x 6 ft.L
Catalog No. 502767335
Encompass_Preferred
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel corrugated inner hose covered by a stainless steel braid reinforcement. The hose has a temperature range of -40°F to 1000°F. It comes in sizes ranging from 1/4 in to 4 in diameter, with lengths from 10 ft to 50 ft. The metal hose is designed for optimal flow while maintaining flexibility and strength. It resists corrosion and abrasion. The hose clamps are included for easy installation. This durable metal hose is ideal for automotive, industrial, and commercial applications.
Catalog No. 50-276-7335 Supplier Grainger Supplier No. 16T783
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel corrugated inner hose covered by a stainless steel braid reinforcement. The hose has a temperature range of -40°F to 1000°F. It comes in sizes ranging from 1/4 in to 4 in diameter, with lengths from 10 ft to 50 ft. The metal hose is designed for optimal flow while maintaining flexibility and strength. It resists corrosion and abrasion. The hose clamps are included for easy installation. This durable metal hose is ideal for automotive, industrial, and commercial applications.