Learn More
Grainger Metal Hose 3 ID x 10 ft.L
Catalog No. 502768324
Encompass_Preferred
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel inner hose covered by a stainless steel wire braid exterior. The hose has a temperature range of -425°F to 1200°F, resists corrosion, abrasion and crushing, and conforms to tight bending radii. It is available in sizes ranging from 1/4 in. to 4 in. inner diameter, with lengths up to 25 ft. The hose fittings have male NPT or socket weld ends.
Catalog No. 50-276-8324 Supplier Grainger Supplier No. 16T844
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a 304 stainless steel inner hose covered by a stainless steel wire braid exterior. The hose has a temperature range of -425°F to 1200°F, resists corrosion, abrasion and crushing, and conforms to tight bending radii. It is available in sizes ranging from 1/4 in. to 4 in. inner diameter, with lengths up to 25 ft. The hose fittings have male NPT or socket weld ends.