Learn More
Grainger Metal Hose 3 ID x 12 ft.L
Catalog No. 502768485
Encompass_Preferred
The Grainger Approved Metal Hose is a 3" ID x 12' L metal hose with a working pressure of 300 psi. It has a zinc-plated steel construction and is designed for use with air, water, oil and other non-flammable fluids. This flexible metal hose features zinc-plated steel wire reinforcement and can withstand temperatures from -40°F to 300°F. Some key specifications include: inside diameter of 3", outside diameter of 4-1/4", length of 12 feet, and weight of 15 lbs. This durable metal hose provides flexibility while maintaining strength and corrosion resistance.
Catalog No. 50-276-8485 Supplier Grainger Supplier No. 16T845
The Grainger Approved Metal Hose is a 3" ID x 12' L metal hose with a working pressure of 300 psi. It has a zinc-plated steel construction and is designed for use with air, water, oil and other non-flammable fluids. This flexible metal hose features zinc-plated steel wire reinforcement and can withstand temperatures from -40°F to 300°F. Some key specifications include: inside diameter of 3", outside diameter of 4-1/4", length of 12 feet, and weight of 15 lbs. This durable metal hose provides flexibility while maintaining strength and corrosion resistance.