Learn More
Grainger Metal Hose 3 ID x 14 ft.L
Catalog No. 502768323
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made from 304 stainless steel with a smooth bore. The hose has a temperature range of -40°F to 1000°F. It has a zinc-plated carbon steel fitting on each end and is 25 feet in length. The hose is designed for use in hydraulic and pneumatic systems to provide flexibility. It resists kinking and abrasion.
Catalog No. 50-276-8323 Supplier Grainger Supplier No. 16T846
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made from 304 stainless steel with a smooth bore. The hose has a temperature range of -40°F to 1000°F. It has a zinc-plated carbon steel fitting on each end and is 25 feet in length. The hose is designed for use in hydraulic and pneumatic systems to provide flexibility. It resists kinking and abrasion.