Learn More
Grainger Metal Hose 3 ID x 18 ft.L
Catalog No. 502766056
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a corrugated interlocked design for flexibility. This metal hose has a temperature range of -40°F to 1000°F and includes a 316 stainless steel braid reinforcement. The fittings are zinc-plated steel fixed male NPT swivel on one end and female NPT swivel on the other end. This durable metal hose is ideal for conveying air, water, oil and other fluids in industrial applications.
Catalog No. 50-276-6056 Supplier Grainger Supplier No. 16T797
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a corrugated interlocked design for flexibility. This metal hose has a temperature range of -40°F to 1000°F and includes a 316 stainless steel braid reinforcement. The fittings are zinc-plated steel fixed male NPT swivel on one end and female NPT swivel on the other end. This durable metal hose is ideal for conveying air, water, oil and other fluids in industrial applications.