Learn More
Grainger Metal Hose 3 ID x 20 ft.L
Catalog No. 502765763
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made from 304 stainless steel with a smooth bore. The hose has a temperature range of -40°F to 1000°F. It has a zinc-plated steel fitting on one end and is 36 inches long when fully extended. The hose allows flexibility while maintaining strength and durability. It can be used for hydraulic lines, lubrication lines, and transfer of water, oil, air or steam. The interlocked construction prevents kinking under pressure.
Catalog No. 50-276-5763 Supplier Grainger Supplier No. 16T798
The Hose Master Interlocked Metal Hose is a 3 inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations and is made from 304 stainless steel with a smooth bore. The hose has a temperature range of -40°F to 1000°F. It has a zinc-plated steel fitting on one end and is 36 inches long when fully extended. The hose allows flexibility while maintaining strength and durability. It can be used for hydraulic lines, lubrication lines, and transfer of water, oil, air or steam. The interlocked construction prevents kinking under pressure.