Learn More
Grainger Metal Hose 3 ID x 6 ft.L
Catalog No. 502766027
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 3 inch diameter hose made from 304 stainless steel. It has a working pressure of 300 PSI and a minimum bend radius of 6 inches. The hose is 25 feet long and has male NPT thread connections on both ends. It can handle temperatures from -40 to 450 degrees Fahrenheit. The interlocked construction allows flexibility while maintaining strength and durability. This metal hose is ideal for conveying air, water, oil and other fluids in industrial applications that require high working pressures and temperatures.
Catalog No. 50-276-6027 Supplier Grainger Supplier No. 16T791
The Hose Master Interlocked Metal Hose is a 3 inch diameter hose made from 304 stainless steel. It has a working pressure of 300 PSI and a minimum bend radius of 6 inches. The hose is 25 feet long and has male NPT thread connections on both ends. It can handle temperatures from -40 to 450 degrees Fahrenheit. The interlocked construction allows flexibility while maintaining strength and durability. This metal hose is ideal for conveying air, water, oil and other fluids in industrial applications that require high working pressures and temperatures.