Learn More
Grainger Metal Hose 4 ID x 12 ft.L
Catalog No. 502766446
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4-inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations for flexibility and is made from 304 stainless steel with a smooth inner wall for optimal flow. This hose meets SAE 100R2 standards and has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT threaded ends for easy attachment. The corrugated stainless steel construction makes this hose very durable while still allowing it to be flexible.
Catalog No. 50-276-6446 Supplier Grainger Supplier No. 16T803
The Hose Master Interlocked Metal Hose is a 4-inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations for flexibility and is made from 304 stainless steel with a smooth inner wall for optimal flow. This hose meets SAE 100R2 standards and has a temperature range of -40°F to 1000°F. It comes in a standard length of 25 feet and has male NPT threaded ends for easy attachment. The corrugated stainless steel construction makes this hose very durable while still allowing it to be flexible.