Learn More
Grainger Metal Hose 4 ID x 14 ft.L
Catalog No. 502765352
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and crush resistance. The hose is made from 304 stainless steel with zinc-plated carbon steel fittings. It has a temperature range of -40°F to 450°F. The hose has a 4-inch NPT male end and 4-inch NPT swivel female end. It is 30 inches long when fully extended. The hose allows movement in all directions while maintaining connections. It resists kinking and has a smooth interior for unrestricted flow. The hose is designed for use transferring air, water, oil and other fluids in industrial applications.
Catalog No. 50-276-5352 Supplier Grainger Supplier No. 16T804
The Hose Master Interlocked Metal Hose is a 4-inch diameter metal hose with a working pressure of 300 PSI. It has interlocked annular corrugations for flexibility and crush resistance. The hose is made from 304 stainless steel with zinc-plated carbon steel fittings. It has a temperature range of -40°F to 450°F. The hose has a 4-inch NPT male end and 4-inch NPT swivel female end. It is 30 inches long when fully extended. The hose allows movement in all directions while maintaining connections. It resists kinking and has a smooth interior for unrestricted flow. The hose is designed for use transferring air, water, oil and other fluids in industrial applications.