Learn More
Grainger Metal Hose 4 ID x 14 ft.L
Catalog No. 502764885
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a corrugated interlocked design for flexibility. This hose has a temperature range of -40 to 450 degrees Fahrenheit. It has a zinc-plated steel fitting on each end and is 30 feet in length. The hose is designed for conveying air, water, oil and other fluids in industrial applications. It meets SAE J517 specifications.
Catalog No. 50-276-4885 Supplier Grainger Supplier No. 16T855
The Hose Master Interlocked Metal Hose is a 4 inch diameter metal hose with a working pressure of 300 PSI. It is made from 304 stainless steel with a corrugated interlocked design for flexibility. This hose has a temperature range of -40 to 450 degrees Fahrenheit. It has a zinc-plated steel fitting on each end and is 30 feet in length. The hose is designed for conveying air, water, oil and other fluids in industrial applications. It meets SAE J517 specifications.